Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002172.2 | NP_002163.2 | 1 | 119 | ||
GenBank mRNA | BC074956.2 | - | 1 | 119 | 30915281 | |
BC104159.1 | - | 1 | 117 | 40029547 | ||
BC104160.1 | - | 1 | 117 | 40029549 | ||
V00542.1 | - | 1 | 131 |