Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plaur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011113.3 | NP_035243.1 | 4 | 429 | ||
GenBank mRNA | AK002580.1 | - | 4 | 500 | ||
AK131951.1 | - | 4 | 445 | |||
AK144796.1 | - | 4 | 446 | |||
AK146827.1 | - | 4 | 444 | |||
AK154141.1 | - | 4 | 446 | |||
AK155351.1 | - | 4 | 433 | |||
BC010309.1 | - | 4 | 429 | |||
X62700.1 | - | 4 | 430 | |||
X62701.1 | - | 4 | 430 |