Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021280.3 | NP_067255.2 | 25 | 2907 | ||
GenBank mRNA | AK134041.1 | - | 24 | 2712 | ||
AK162809.1 | - | 13 | 1579 | |||
BC057161.1 | - | 25 | 2922 | |||
BC065091.1 | - | 25 | 2907 | 6854923 | ||
X95346.1 | - | 3 | 238 |