Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
Plec
Gene Name:
plectin
Gene Aliases:
AA591047, AU042537, EBS1, PCN, PLTN, Plec1
Chromosome Location:
Chr.15: 76170974 - 76231378 on Build GRCm38
UniGene ID:
Mm.234912
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s71823
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Plec
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163540.1
NP_001157012.1
7
1203
NM_001163542.1
NP_001157014.1
8
720
NM_001163549.1
NP_001157021.1
7
766
NM_001164203.1
NP_001157675.1
8
707
NM_011117.2
NP_035247.2
6
669
NM_201385.2
NP_958787.2
6
604
NM_201386.2
NP_958788.2
6
665
NM_201387.3
NP_958789.3
6
694
NM_201388.2
NP_958790.2
6
656
NM_201389.2
NP_958791.2
6
1188
NM_201390.2
NP_958792.2
6
603
NM_201391.2
NP_958793.2
6
751
NM_201392.2
NP_958794.2
6
577
NM_201393.2
NP_958795.2
6
694
NM_201394.2
NP_958796.2
6
751
GenBank mRNA
AF188008.1
-
7
1203
AF188011.1
-
7
766
AF188012.1
-
6
751
AF188013.1
-
8
720
AF188014.1
-
6
669
AF188021.1
-
6
577
AF188023.1
-
8
707
AK017743.1
-
6
584
AK044559.1
-
764
AY480033.1
-
6
669
AY480034.1
-
6
604
AY480035.1
-
6
665
AY480036.1
-
6
694
AY480037.1
-
6
656
AY480038.1
-
6
1188
AY480039.1
-
6
603
AY480040.1
-
6
751
AY480041.1
-
6
577
AY480042.1
-
6
694
AY480043.1
-
6
751
BC021810.1
-
6
738
BC099929.1
-
6
1189
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
response to nutrient
hemidesmosome assembly
actin binding
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline