Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011121.3 | NP_035251.3 | 9 | 1562 | ||
GenBank mRNA | AK134045.1 | - | 9 | 1503 | ||
AK148050.1 | - | 9 | 1476 | |||
AK148076.1 | - | 9 | 1553 | |||
AK164027.1 | - | 9 | 1532 | |||
BC006880.1 | - | 9 | 1514 | 3596381 | ||
L19558.1 | - | 9 | 1569 | |||
U01063.1 | - | 9 | 1483 |