Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011122.3 | NP_035252.1 | 9 | 1044 | ||
GenBank mRNA | AF046782.1 | - | 9 | 972 | ||
AK137480.1 | - | 9 | 994 | |||
AK157729.1 | - | 9 | 994 | |||
AK160822.1 | - | 9 | 1011 | |||
BC006599.1 | - | 9 | 969 | 3965301 |