Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011125.2 | NP_035255.1 | 12, 13 | 1309 | ||
GenBank mRNA | AK148330.1 | - | 12, 13 | 1314 | ||
BC003782.1 | - | 12, 13 | 1243 | 3491360 | ||
CT010317.1 | - | 11, 12 | 1173 | |||
U28960.1 | - | 11, 12 | 1167 | |||
U37226.1 | - | 12, 13 | 1239 |