Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011125.2 | NP_035255.1 | 5 | 543 | ||
GenBank mRNA | AK148330.1 | - | 5 | 548 | ||
AK149730.1 | - | 5 | 474 | |||
AK190857.1 | - | 2 | 207 | |||
U28960.1 | - | 4 | 401 | |||
U37226.1 | - | 5 | 473 |