Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011125.2 | NP_035255.1 | 4, 5 | 457 | ||
GenBank mRNA | AK148330.1 | - | 4, 5 | 462 | ||
AK149730.1 | - | 4, 5 | 388 | |||
AK190857.1 | - | 1, 2 | 121 | |||
BC003782.1 | - | 4, 5 | 391 | 3491360 | ||
CT010317.1 | - | 3, 4 | 321 | |||
U37226.1 | - | 4, 5 | 387 |