Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002176.3 | NP_002167.1 | 1 | 331 | ||
GenBank mRNA | AB451323.1 | - | 1 | 237 | ||
AB451452.1 | - | 1 | 237 | |||
AB451491.1 | - | 1 | 174 | |||
BC069314.1 | - | 1 | 237 | 7262165 | ||
BC096150.3 | - | 1 | 237 | 40002940 | ||
BC096151.2 | - | 1 | 237 | 40002941 | ||
BC096152.2 | - | 1 | 237 | 40002943 | ||
BC096153.2 | - | 1 | 237 | 40002945 | ||
M28622.1 | - | 1 | 312 | |||
V00546.1 | - | 1 | 301 | |||
V00547.1 | - | 1 | 237 |