Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008881.2 | NP_032907.1 | 27 | 5132 | ||
GenBank mRNA | AK154544.1 | - | 10 | 1542 | ||
AK220460.1 | - | 24 | 3372 | |||
BC138023.1 | - | 26 | 4924 | 8861042 | ||
D86948.1 | - | 27 | 5116 |