Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008884.5 | NP_032910.3 | 3 | 797 | ||
NM_178087.4 | NP_835188.2 | 3 | 797 | |||
GenBank mRNA | AK028044.1 | - | 3 | 799 | ||
AK052305.1 | - | 3 | 797 | |||
AK054135.1 | - | 3 | 797 | |||
AK137538.1 | - | 3 | 745 | |||
AK156279.1 | - | 3 | 679 | |||
BC020990.1 | - | 3 | 742 | 4188386 | ||
KJ650238.1 | - | 3 | 701 | |||
U33626.2 | - | 3 | 694 |