Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076005.1 | - | 4 | 421 | ||
AK171745.1 | - | 4 | 418 | |||
BC010765.1 | - | 4 | 444 | 4225316 | ||
M32240.1 | - | 4 | 471 | |||
Z38110.1 | - | 3 | 285 |