Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008886.2 | NP_032912.2 | 13 | 2257 | ||
GenBank mRNA | AK083049.1 | - | 13 | 2214 | ||
AK146365.1 | - | 13 | 2270 | |||
BC022179.1 | - | 12 | 2057 | |||
BC058189.1 | - | 13 | 2263 | |||
BC066110.1 | - | 13 | 2257 | |||
BC083502.1 | - | 13 | 2217 | |||
BC086637.1 | - | 13 | 2233 | |||
BC110684.1 | - | 12 | 2885 | |||
BC139774.1 | - | 13 | 1691 | |||
BC141287.1 | - | 13 | 2233 | 9055899 |