Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008889.2 | NP_032915.2 | 2 | 494 | ||
GenBank mRNA | AK141152.1 | - | 2 | 575 | ||
AK165792.1 | - | 2 | 108 | |||
BC082545.1 | - | 2 | 494 | 6820968 | ||
BC125650.1 | - | 2 | 377 | 40130165 | ||
BC125654.1 | - | 2 | 391 | 40130169 | ||
CF739461.1 | - | 2 | 98 | |||
X97490.1 | - | 2 | 557 |