Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011130.2 | NP_035260.1 | 8, 9 | 548 | ||
GenBank mRNA | AF513911.1 | - | 8, 9 | 525 | ||
AK077127.1 | - | 8, 9 | 533 | |||
AK146745.1 | - | 8, 9 | 537 | |||
AK151436.1 | - | 8, 9 | 533 | |||
BC006681.1 | - | 7, 8 | 496 | |||
BC060998.1 | - | 8, 9 | 529 | 6774075 |