Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008894.2 | NP_032920.2 | 8, 9 | 1134 | ||
GenBank mRNA | AK013901.1 | - | 8, 9 | 1100 | ||
BC003342.1 | - | 8, 9 | 1100 | 3257610 | ||
CB205181.1 | - | 5, 6 | 670 | |||
Z72486.1 | - | 8, 9 | 1095 |