Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011134.3 | NP_035264.2 | 1 | 262 | ||
GenBank mRNA | AK050119.1 | - | 1 | 80 | ||
BB870662.1 | - | 1 | 263 | |||
BC012706.1 | - | 1 | 135 | 4158951 | ||
U32684.1 | - | 1 | 144 | |||
U72636.1 | - | 1 | 144 |