Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271542.1 | NP_001258471.1 | 4 | 597 | ||
NM_001271543.1 | NP_001258472.1 | 4 | 668 | |||
NM_011135.5 | NP_035265.1 | 4 | 668 | |||
GenBank mRNA | AK044535.1 | - | 3 | 1534 | ||
AK132063.1 | - | 4 | 588 | |||
AK135669.1 | - | 4 | 599 | |||
AK137838.1 | - | 4 | 633 | |||
AK147048.1 | - | 4 | 635 | |||
AK166449.1 | - | 4 | 632 | |||
BC006021.1 | - | 4 | 612 | 3590764 |