Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011137.3 | NP_035267.2 | 3 | 157 | ||
NM_198932.3 | NP_945150.2 | 3 | 157 | |||
NM_198933.3 | NP_945151.2 | 3 | 229 | |||
NM_198934.3 | NP_945152.2 | 3 | 229 | |||
GenBank mRNA | AF508939.1 | - | 3 | 138 | ||
AJ296212.1 | - | 3 | 181 | |||
AJ489474.1 | - | 3 | 229 | |||
AJ489475.1 | - | 3 | 220 | |||
AK028237.1 | - | 3 | 159 | |||
AY177625.1 | - | 3 | 138 | |||
BC066201.1 | - | 3 | 133 | |||
S65461.1 | - | 2 | 57 | |||
X56230.1 | - | 3 | 288 | |||
X68362.1 | - | 3 | 130 | |||
X68363.1 | - | 3 | 128 | |||
X68364.1 | - | 3 | 144 |