Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou4f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138944.2 | NP_620394.2 | 2 | 1005 | ||
GenBank mRNA | AK084240.1 | - | 2 | 1004 | ||
AK087546.1 | - | 2 | 1005 | |||
S68377.1 | - | 2 | 988 |