Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163557.1 | NP_001157029.1 | 11, 12 | 1194 | ||
NM_008905.2 | NP_032931.2 | 11, 12 | 1430 | |||
GenBank mRNA | AK012152.1 | - | 8, 9 | 853 | ||
BC055935.1 | - | 1, 2 | 64 | |||
BC100346.1 | - | 11, 12 | 1194 | 30936010 | ||
U79024.1 | - | 11, 12 | 1215 |