Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024459.2 | NP_077779.2 | 5 | 607 | ||
GenBank mRNA | AK133403.1 | - | 578 | |||
AK145541.1 | - | 5 | 584 | |||
AK160423.1 | - | 552 | ||||
AK160938.1 | - | 5 | 593 | |||
AK163241.1 | - | 5 | 581 | |||
BC002267.1 | - | 3 | 435 | |||
S43864.1 | - | 4 | 365 |