Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293650.1 | NP_001280579.1 | 13 | 1310 | ||
NM_001293651.1 | NP_001280580.1 | 13 | 1293 | |||
NM_008925.2 | NP_032951.1 | 13 | 1289 | |||
GenBank mRNA | AK137661.1 | - | 13 | 1256 | ||
AK153931.1 | - | 13 | 1264 | |||
AK159705.1 | - | 13 | 1260 | |||
AK177012.1 | - | 2 | 80 | |||
AK208791.1 | - | 1 | 30 | |||
BC009816.1 | - | 13 | 1243 | 4008182 | ||
CX207653.1 | - | 5 | 429 | |||
U92794.1 | - | 13 | 1242 |