Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016979.1 | NP_058675.1 | 5 | 1001 | ||
GenBank mRNA | AJ238004.1 | - | 5 | 884 | ||
AK034681.1 | - | 5 | 1320 | |||
AK037141.1 | - | 5 | 995 | |||
AK081548.1 | - | 5 | 981 | |||
AK139510.1 | - | 5 | 1009 | |||
AK149937.1 | - | 5 | 979 | |||
AK150588.1 | - | 5 | 963 | |||
AK154447.1 | - | 5 | 981 | |||
AK169322.1 | - | 5 | 952 | |||
BC006875.1 | - | 5 | 1001 | 3596013 |