Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271378.1 | NP_001258307.1 | 3, 4 | 351 | ||
NM_011167.3 | NP_035297.1 | 3, 4 | 348 | |||
GenBank mRNA | AF090140.1 | - | 3, 4 | 257 | ||
AK005435.1 | - | 3, 4 | 312 | |||
BC099469.1 | - | 3, 4 | 294 | 30787452 |