Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011169.5 | NP_035299.4 | 10 | 1779 | ||
GenBank mRNA | AK054551.1 | - | 9 | 2290 | ||
BC005555.1 | - | 10 | 1362 | 3586308 | ||
BC006652.1 | - | 10 | 1253 | 3500076 | ||
D10214.1 | - | 8 | 1054 | |||
L13593.1 | - | 8 | 1089 | |||
L14811.1 | - | 8 | 1031 | |||
X73372.1 | - | 8 | 1031 |