Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013637.4 | NP_038665.1 | 2 | 346 | ||
GenBank mRNA | AK006377.1 | - | 2 | 345 | ||
AK007068.1 | - | 2 | 346 | |||
AK018980.1 | - | 2 | 349 | |||
BC049599.1 | - | 345 | 6773822 | |||
BC059733.1 | - | 2 | 345 | 6774637 |