Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008941.3 | NP_032967.1 | 12, 13 | 1331 | ||
NM_178855.4 | NP_849186.2 | 11, 12 | 1286 | |||
GenBank mRNA | AK033582.1 | - | 11, 12 | 1288 | ||
AK038356.1 | - | 3, 4 | 280 | |||
BC117917.1 | - | 12, 13 | 1330 | 40097367 | ||
BC117918.1 | - | 12, 13 | 1330 | 40097370 | ||
U73378.1 | - | 12, 13 | 1326 |