Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146120.1 | NP_001139592.1 | 11 | 1471 | ||
NM_001146121.1 | NP_001139593.1 | 12 | 1477 | |||
NM_001146122.1 | NP_001139594.1 | 11 | 1468 | |||
NM_001146123.1 | NP_001139595.1 | 11 | 1444 | |||
NM_001146124.1 | NP_001139596.1 | 11 | 1387 | |||
NM_011179.3 | NP_035309.3 | 12 | 1480 | |||
GenBank mRNA | AK088369.1 | - | 11 | 1385 | ||
AK089998.1 | - | 11 | 1389 | |||
AK093881.1 | - | 11 | 1385 | |||
AK135637.1 | - | 11 | 1385 | |||
AK145607.1 | - | 11 | 1392 | |||
AK145802.1 | - | 11 | 1386 | |||
AK148150.1 | - | 11 | 1393 | |||
AK148152.1 | - | 11 | 1412 | |||
AK148545.1 | - | 12 | 1396 | |||
AK149785.1 | - | 11 | 1388 | |||
AK150331.1 | - | 11 | 1387 | |||
AK151034.1 | - | 11 | 1385 | |||
AK151173.1 | - | 11 | 1386 | |||
AK151230.1 | - | 11 | 1385 | |||
AK151234.1 | - | 11 | 1360 | |||
AK151251.1 | - | 11 | 1358 | |||
AK151525.1 | - | 11 | 1387 | |||
AK151529.1 | - | 11 | 1360 | |||
AK151686.1 | - | 11 | 1421 | |||
AK151852.1 | - | 11 | 1387 | |||
AK151875.1 | - | 11 | 1384 | |||
AK151891.1 | - | 11 | 1385 | |||
AK151907.1 | - | 11 | 1387 | |||
AK151943.1 | - | 11 | 1387 | |||
AK152117.1 | - | 11 | 1387 | |||
AK152126.1 | - | 11 | 1385 | |||
AK152134.1 | - | 11 | 1387 | |||
AK152203.1 | - | 11 | 1419 | |||
AK152280.1 | - | 11 | 1385 | |||
AK152319.1 | - | 11 | 1389 | |||
AK152355.1 | - | 11 | 1387 | |||
AK152408.1 | - | 11 | 1390 | |||
AK152505.1 | - | 11 | 1387 | |||
AK152557.1 | - | 11 | 1387 | |||
AK152605.1 | - | 11 | 1387 | |||
AK152819.1 | - | 11 | 1399 | |||
AK153404.1 | - | 11 | 1385 | |||
AK159108.1 | - | 11 | 1415 | |||
AK159164.1 | - | 11 | 1385 | |||
AK159167.1 | - | 11 | 1386 | |||
AK159198.1 | - | 11 | 1385 | |||
AK159221.1 | - | 11 | 1386 | |||
AK159244.1 | - | 11 | 1382 | |||
AK159266.1 | - | 11 | 1385 | |||
AK159333.1 | - | 11 | 1385 | |||
AK159341.1 | - | 11 | 1385 | |||
AK159384.1 | - | 11 | 1385 | |||
AK159414.1 | - | 11 | 1388 | |||
AK159427.1 | - | 11 | 1387 | |||
AK159451.1 | - | 11 | 1385 | |||
AK159457.1 | - | 11 | 1399 | |||
AK159463.1 | - | 11 | 1387 | |||
AK159487.1 | - | 11 | 1385 | |||
AK159492.1 | - | 11 | 1382 | |||
AK159510.1 | - | 11 | 1385 | |||
AK159528.1 | - | 11 | 1385 | |||
AK159530.1 | - | 11 | 1385 | |||
AK159589.1 | - | 11 | 1387 | |||
AK159604.1 | - | 11 | 1385 | |||
AK159622.1 | - | 10 | 1277 | |||
AK159634.1 | - | 11 | 1385 | |||
AK159638.1 | - | 11 | 1386 | |||
AK159657.1 | - | 11 | 1385 | |||
AK159664.1 | - | 11 | 1385 | |||
AK159685.1 | - | 11 | 1385 | |||
AK159692.1 | - | 11 | 1385 | |||
AK159709.1 | - | 11 | 1385 | |||
AK159720.1 | - | 11 | 1385 | |||
AK159722.1 | - | 11 | 1385 | |||
AK159728.1 | - | 11 | 1387 | |||
AK159840.1 | - | 11 | 1385 | |||
AK159861.1 | - | 11 | 1385 | |||
AK159862.1 | - | 11 | 1387 | |||
AK159877.1 | - | 11 | 1385 | |||
AK159910.1 | - | 11 | 1385 | |||
AK159915.1 | - | 11 | 1385 | |||
AK159936.1 | - | 11 | 1387 | |||
AK159982.1 | - | 11 | 1385 | |||
AK159994.1 | - | 11 | 1385 | |||
AK160015.1 | - | 11 | 1385 | |||
AK160019.1 | - | 11 | 1385 | |||
AK160021.1 | - | 11 | 1383 | |||
AK160024.1 | - | 11 | 1385 | |||
AK160046.1 | - | 11 | 1385 | |||
AK160049.1 | - | 11 | 1385 | |||
AK160073.1 | - | 11 | 1383 | |||
AK160108.1 | - | 11 | 1383 | |||
AK160877.1 | - | 11 | 1388 | |||
AK161313.1 | - | 11 | 1402 | |||
AK166700.1 | - | 11 | 1385 | |||
AK166707.1 | - | 11 | 1384 | |||
AK166744.1 | - | 11 | 1383 | |||
AK166811.1 | - | 11 | 1385 | |||
AK166849.1 | - | 11 | 1385 | |||
AK166879.1 | - | 11 | 1387 | |||
AK166909.1 | - | 11 | 1385 | |||
AK166920.1 | - | 11 | 1385 | |||
AK166946.1 | - | 11 | 1392 | |||
AK166959.1 | - | 11 | 1385 | |||
AK166984.1 | - | 11 | 1387 | |||
AK167014.1 | - | 11 | 1385 | |||
AK167025.1 | - | 11 | 1385 | |||
AK167073.1 | - | 11 | 1385 | |||
AK167099.1 | - | 11 | 1390 | |||
AK167108.1 | - | 11 | 1387 | |||
AK167111.1 | - | 11 | 1385 | |||
AK167147.1 | - | 11 | 1385 | |||
AK167198.1 | - | 11 | 1392 | |||
AK167213.1 | - | 11 | 1385 | |||
AK167237.1 | - | 11 | 1385 | |||
AK167243.1 | - | 11 | 1388 | |||
AK167252.1 | - | 11 | 1389 | |||
AK167418.1 | - | 11 | 1387 | |||
AK167719.1 | - | 11 | 1387 | |||
AK167902.1 | - | 11 | 1455 | |||
AK170416.1 | - | 11 | 1384 | |||
AK182220.1 | - | 2 | 157 | |||
AK206110.1 | - | 1 | 76 | |||
BC036266.1 | - | 10 | 1251 | |||
BC141089.1 | - | 12 | 1390 | 9055701 | ||
BC145359.1 | - | 11 | 1384 | 9053917 | ||
S36200.1 | - | 12 | 1370 | |||
S71616.1 | - | 12 | 1410 | |||
U27340.1 | - | 11 | 1385 |