Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112699.1 | NP_001106169.1 | 12 | 1115 | ||
NM_001112700.1 | NP_001106170.1 | 13 | 1058 | |||
NM_011180.3 | NP_035310.2 | 13 | 1118 | |||
GenBank mRNA | AB013464.1 | - | 12 | 1036 | ||
AB035538.1 | - | 12 | 1087 | |||
AF051337.1 | - | 12 | 986 | |||
AK078502.1 | - | 1025 | ||||
AK158216.1 | - | 13 | 1058 | |||
AK172296.1 | - | 13 | 1063 | |||
AK220302.1 | - | 13 | 1040 | |||
AY126516.1 | - | 12 | 1084 | |||
BC057974.1 | - | 12 | 999 | 5060913 |