Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112699.1 | NP_001106169.1 | 3 | 266 | ||
NM_001112700.1 | NP_001106170.1 | 3 | 206 | |||
NM_011180.3 | NP_035310.2 | 3 | 266 | |||
GenBank mRNA | AB013464.1 | - | 3 | 187 | ||
AB035538.1 | - | 3 | 235 | |||
AF051337.1 | - | 2 | 134 | |||
AK078502.1 | - | 177 | ||||
AK158216.1 | - | 3 | 206 | |||
AK172296.1 | - | 3 | 211 | |||
AK220302.1 | - | 3 | 188 | |||
AY126516.1 | - | 3 | 235 | |||
BC057974.1 | - | 3 | 150 | 5060913 | ||
BY204859.1 | - | 3 | 206 | |||
CJ131185.1 | - | 3 | 266 |