Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112701.1 | NP_001106171.1 | 7 | 799 | ||
NM_011181.3 | NP_035311.1 | 7 | 799 | |||
GenBank mRNA | AB013466.1 | - | 7 | 734 | ||
AB013467.1 | - | 6 | 1302 | |||
AF079971.1 | - | 7 | 768 | |||
AK045451.1 | - | 7 | 768 | |||
AK143829.1 | - | 7 | 741 | |||
AK168979.1 | - | 7 | 739 | |||
AK172050.1 | - | 7 | 755 | |||
BC004662.1 | - | 7 | 713 | 3498490 | ||
BC088833.1 | - | 7 | 773 | 5293465 |