Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163548.1 | NP_001157020.1 | 8, 9 | 1231 | ||
NM_011182.4 | NP_035312.3 | 8, 9 | 907 | |||
GenBank mRNA | AB013470.1 | - | 8, 9 | 761 | ||
AF084221.1 | - | 8, 9 | 801 | |||
AK016645.1 | - | 8, 9 | 1233 | |||
AK036191.1 | - | 8, 9 | 821 | |||
AK142407.1 | - | 8, 9 | 896 | |||
AK159229.1 | - | 8, 9 | 879 | |||
AK168566.1 | - | 8, 9 | 821 | |||
AK184003.1 | - | 1, 2 | 91 | |||
AK186497.1 | - | 1, 2 | 91 | |||
AK220565.1 | - | 8, 9 | 887 | |||
BC007189.1 | - | 7, 8 | 648 | |||
BC035296.1 | - | 8, 9 | 848 | 3969080 |