Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128605.1 | NP_001122077.1 | 4 | 556 | ||
NM_011183.3 | NP_035313.2 | 4 | 565 | |||
GenBank mRNA | AK014706.1 | - | 3 | 500 | ||
AK148111.1 | - | 4 | 557 | |||
AK154119.1 | - | 4 | 501 | |||
BC010403.1 | - | 3 | 511 | 4038761 | ||
U57324.1 | - | 4 | 516 |