Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008947.3 | NP_032973.1 | 8 | 1007 | ||
GenBank mRNA | AK088411.1 | - | 9 | 1037 | ||
AK133926.1 | - | 9 | 1041 | |||
AK148906.1 | - | 9 | 1029 | |||
AK161641.1 | - | 9 | 1056 | |||
AK161723.1 | - | 9 | 1057 | |||
AK166783.1 | - | 9 | 1022 | |||
AK169015.1 | - | 9 | 1020 | |||
BC003860.1 | - | 9 | 1023 | 3486436 | ||
U39302.1 | - | 8 | 1008 |