Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008947.3 | NP_032973.1 | 8 | 934 | ||
GenBank mRNA | AK088411.1 | - | 9 | 964 | ||
AK161641.1 | - | 9 | 983 | |||
AK161723.1 | - | 9 | 984 | |||
AK166783.1 | - | 9 | 949 | |||
AK169015.1 | - | 9 | 947 | |||
BC003860.1 | - | 9 | 950 | 3486436 | ||
U39302.1 | - | 8 | 935 |