Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008948.2 | NP_032974.2 | 8, 9 | 1134 | ||
GenBank mRNA | AK002542.1 | - | 8, 9 | 855 | ||
AK083043.1 | - | 1096 | ||||
AK150189.1 | - | 8, 9 | 1070 | |||
AK151370.1 | - | 8, 9 | 854 | |||
AK152018.1 | - | 8, 9 | 873 | |||
AK167002.1 | - | 8, 9 | 1082 | |||
AK167061.1 | - | 8, 9 | 1079 | |||
AK168263.1 | - | 7, 8 | 803 | |||
AK168616.1 | - | 8, 9 | 854 | |||
BC005783.1 | - | 8, 9 | 1023 | 3603053 | ||
D49686.1 | - | 8, 9 | 1078 |