Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008950.1 | NP_032976.1 | 5 | 384 | ||
GenBank mRNA | AK010505.1 | - | 5 | 404 | ||
AK135451.1 | - | 5 | 394 | |||
AK145703.1 | - | 5 | 388 | |||
BC004052.1 | - | 1 | 79 | |||
BC030840.1 | - | 5 | 416 | 3994523 | ||
Z54219.1 | - | 5 | 384 |