Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008950.1 | NP_032976.1 | 8 | 894 | ||
GenBank mRNA | AK010505.1 | - | 8 | 914 | ||
AK012294.1 | - | 8 | 927 | |||
AK135451.1 | - | 8 | 904 | |||
AK145703.1 | - | 8 | 898 | |||
BC004052.1 | - | 4 | 589 | |||
BC030840.1 | - | 8 | 926 | 3994523 | ||
Z54219.1 | - | 8 | 894 |