Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282017.1 | NP_001268946.1 | 2 | 184 | ||
NM_008951.2 | NP_032977.1 | 2 | 184 | |||
GenBank mRNA | AB029142.1 | - | 2 | 115 | ||
AB029143.1 | - | 2 | 115 | |||
AB029144.1 | - | 2 | 113 | |||
AB029145.1 | - | 2 | 115 | |||
AB029146.1 | - | 2 | 103 | |||
AF013099.1 | - | 2 | 115 | |||
AF175574.1 | - | 2 | 115 | |||
AK010969.1 | - | 2 | 113 | |||
AK145524.1 | - | 2 | 123 | |||
BC009005.1 | - | 2 | 133 | 3581937 | ||
BC016433.1 | - | 2 | 118 | |||
BY099585.1 | - | 2 | 185 |