Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029855.1 | NP_001025026.1 | 3 | 152 | ||
NM_011190.3 | NP_035320.1 | 3 | 252 | |||
GenBank mRNA | AA267668.1 | - | 3 | 143 | ||
AK012344.1 | - | 3 | 181 | |||
AK088654.1 | - | 3 | 179 | |||
AK143556.1 | - | 3 | 184 | |||
BB869062.1 | - | 3 | 152 | |||
BC005680.1 | - | 3 | 172 | 3154255 | ||
BC057859.1 | - | 3 | 249 | 5310526 | ||
BU563130.1 | - | 3 | 321 | |||
BY134400.1 | - | 3 | 218 | |||
CA536627.1 | - | 3 | 164 | |||
U60329.1 | - | 3 | 121 |
Gene Symbol: Psme2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281472.1 | NP_001268401.1 | 1 | 412 | ||
GenBank mRNA | BE624310.1 | - | 1 | 412 | ||
D87910.1 | - | 1 | 139 |