Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pipox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008952.2 | NP_032978.2 | 6 | 1000 | ||
GenBank mRNA | AK008555.2 | - | 6 | 961 | ||
AK149488.1 | - | 6 | 949 | |||
BC013525.1 | - | 6 | 914 | 4237443 | ||
U94700.1 | - | 6 | 912 |