Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011193.2 | NP_035323.2 | 14 | 1500 | ||
GenBank mRNA | AK037936.1 | - | 12 | 2491 | ||
AK088150.1 | - | 13 | 2302 | |||
BC096761.1 | - | 14 | 1280 | 30040955 | ||
U87814.1 | - | 14 | 1340 |