Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077363.2 | NP_001070831.1 | 4 | 413 | ||
NM_008956.3 | NP_032982.2 | 4 | 413 | |||
GenBank mRNA | AB074753.1 | - | 3 | 131 | ||
AB074764.1 | - | 3 | 131 | |||
AK033319.1 | - | 4 | 181 | |||
AK037607.1 | - | 4 | 213 | |||
AK041143.1 | - | 4 | 181 | |||
AK043824.1 | - | 4 | 206 | |||
AK044956.1 | - | 4 | 209 | |||
AK050498.1 | - | 4 | 178 | |||
AK051669.1 | - | 4 | 181 | |||
AK053926.1 | - | 4 | 178 | |||
AK082600.1 | - | 4 | 217 | |||
AK088126.1 | - | 4 | 181 | |||
AK088483.1 | - | 4 | 178 | |||
AK149941.1 | - | 4 | 178 | |||
AK150840.1 | - | 4 | 217 | |||
AK151292.1 | - | 4 | 241 | |||
AK153561.1 | - | 4 | 241 | |||
AK163274.1 | - | 4 | 238 | |||
AK168359.1 | - | 4 | 178 | |||
AK172712.1 | - | 4 | 213 | |||
AK181227.1 | - | 2 | 67 | |||
AK181522.1 | - | 2 | 67 | |||
AK187795.1 | - | 2 | 68 | |||
AK202631.1 | - | 2 | 80 | |||
AK205920.1 | - | 2 | 67 | |||
AK207638.1 | - | 2 | 73 | |||
AK214271.1 | - | 2 | 67 | |||
AK216603.1 | - | 2 | 67 | |||
AK219933.1 | - | 2 | 67 | |||
AK220042.1 | - | 1 | 20 | |||
BC007472.1 | - | 4 | 230 | 3601527 | ||
BC028848.1 | - | 4 | 180 | 4022742 | ||
BC066210.1 | - | 4 | 190 | 30010932 | ||
BC086489.1 | - | 4 | 165 | 30439895 | ||
BY167466.1 | - | 4 | 241 | |||
X52101.1 | - | 4 | 165 |