Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077363.2 | NP_001070831.1 | 12 | 1576 | ||
NM_001283013.1 | NP_001269942.1 | 11 | 1378 | |||
NM_008956.3 | NP_032982.2 | 11 | 1498 | |||
GenBank mRNA | AB074753.1 | - | 10 | 1216 | ||
AB074764.1 | - | 11 | 1294 | |||
AK033319.1 | - | 12 | 1344 | |||
AK036745.1 | - | 11 | 1143 | |||
AK037607.1 | - | 9 | 1341 | |||
AK041143.1 | - | 12 | 1344 | |||
AK043824.1 | - | 12 | 1369 | |||
AK044956.1 | - | 11 | 1294 | |||
AK050498.1 | - | 12 | 1341 | |||
AK051669.1 | - | 11 | 1266 | |||
AK053926.1 | - | 12 | 1342 | |||
AK082600.1 | - | 11 | 1302 | |||
AK088126.1 | - | 9 | 998 | |||
AK088483.1 | - | 12 | 1341 | |||
AK149941.1 | - | 11 | 1263 | |||
AK150840.1 | - | 11 | 1302 | |||
AK151292.1 | - | 12 | 1404 | |||
AK153561.1 | - | 12 | 1404 | |||
AK168359.1 | - | 11 | 1263 | |||
AK168392.1 | - | 11 | 1305 | |||
AK172712.1 | - | 11 | 1301 | |||
AK206119.1 | - | 4 | 359 | |||
BC007472.1 | - | 12 | 1393 | 3601527 | ||
BC008531.1 | - | 8 | 821 | |||
BC028848.1 | - | 12 | 1343 | 4022742 | ||
BC066210.1 | - | 12 | 1353 | 30010932 | ||
BC086489.1 | - | 12 | 1328 | 30439895 | ||
X52101.1 | - | 11 | 1248 |