Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008957.2 | NP_032983.1 | 2 | 298 | ||
GenBank mRNA | AB164615.1 | - | 2 | 214 | ||
AB164616.1 | - | 2 | 174 | |||
AB189441.1 | - | 2 | 214 | |||
AB189442.1 | - | 2 | 174 | |||
AK147626.1 | - | 2 | 508 | |||
U46155.1 | - | 2 | 298 |