Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008958.2 | NP_032984.1 | 20, 21 | 3257 | ||
GenBank mRNA | AB010833.1 | - | 20, 21 | 3257 | ||
AK042653.1 | - | 731 | ||||
BC058397.1 | - | 20, 21 | 3512 |