Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008963.2 | NP_032989.2 | 2, 3 | 327 | ||
GenBank mRNA | AB006361.1 | - | 2, 3 | 310 | ||
AK002382.1 | - | 1, 2 | 152 | |||
AK131859.1 | - | 2, 3 | 327 | |||
BC038083.1 | - | 1, 2 | 403 | 4481308 | ||
BC043015.1 | - | 2, 3 | 313 | 5683706 | ||
X89222.1 | - | 2, 3 | 298 |