Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptger1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013641.2 | NP_038669.1 | 2 | 1180 | ||
GenBank mRNA | AK053616.1 | - | 954 | |||
AK164746.1 | - | 988 | ||||
BC138739.1 | - | 2 | 1142 | 8861761 | ||
BC138744.1 | - | 2 | 1142 | 8861766 | ||
D16338.1 | - | 2 | 957 |
Set Membership: |
G-protein coupled receptor |